Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem41b
Entrez Gene ID
361626
Gene Name
transmembrane protein 41B
Gene Aliases
RGD1310870
Chromosome Location
Chr.1: 174605833 - 174620185 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012358.1 NP_001012358.1 4 - 5 583 60
GenBank mRNA BC089866.1 - 4 - 5 583 60

View Full Product Details