Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zswim2
Entrez Gene ID
296455
Gene Name
zinc finger, SWIM-type containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 71274341 - 71295644 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011960.1 NP_001011960.1 7 - 8 976 83
XM_006234435.2 XP_006234497.1 7 - 8 1010 83
GenBank mRNA BC079431.1 - 7 - 8 976 83

View Full Product Details