Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phactr4
Entrez Gene ID
682920
Gene Name
phosphatase and actin regulator 4
Gene Aliases
-
Chromosome Location
Chr.5: 150533895 - 150609422 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764195.1 XP_008762417.1 14 - 15 2702 82

View Full Product Details