Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bsdc1
Entrez Gene ID
297890
Gene Name
BSD domain containing 1
Gene Aliases
RGD1311622
Chromosome Location
Chr.5: 147661618 - 147689766 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106636.1 NP_001100106.1 9 - 10 1171 60

View Full Product Details