Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ormdl2
Entrez Gene ID
288783
Gene Name
ORMDL sphingolipid biosynthesis regulator 2
Gene Aliases
-
Chromosome Location
Chr.7: 3242991 - 3246097 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105940.1 NP_001099410.1 2 - 3 382 69
XM_006240753.2 XP_006240815.1 2 - 3 362 69
XM_006240754.2 XP_006240816.1 2 - 3 282 69

View Full Product Details