Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cd63
Entrez Gene ID
29186
Gene Name
Cd63 molecule
Gene Aliases
-
Chromosome Location
Chr.7: 3320250 - 3335581 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017125.3 NP_058821.1 7 - 8 717 58
XM_008765024.1 XP_008763246.1 7 - 8 779 58
XM_008765025.1 XP_008763247.1 7 - 8 982 58
GenBank mRNA BC063173.1 - 7 - 8 720 58
FQ214468.1 - 7 - 8 784 58
FQ221333.1 - 7 - 8 721 58
FQ222038.1 - 7 - 8 700 58
FQ230116.1 - 7 - 8 688 58
X61654.1 - 7 - 8 717 58

View Full Product Details