The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107694.1 | NP_001101164.1 | 10 - 11 | 986 | 63 |
XM_006232876.2 | XP_006232938.1 | 9 - 10 | 2484 | 63 | |
XM_006232878.2 | XP_006232940.1 | 9 - 10 | 2793 | 63 | |
XM_008761300.1 | XP_008759522.1 | 8 - 9 | 1235 | 63 | |
XM_008761301.1 | XP_008759523.1 | 9 - 10 | 2739 | 63 |