Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Man1a2
Entrez Gene ID
295319
Gene Name
mannosidase, alpha, class 1A, member 2
Gene Aliases
Man1b
Chromosome Location
Chr.2: 202891537 - 203043847 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106452.1 NP_001099922.1 1 - 2 665 83

View Full Product Details