Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pth2r
Entrez Gene ID
81753
Gene Name
parathyroid hormone 2 receptor
Gene Aliases
Pthr2
Chromosome Location
Chr.9: 72052966 - 72157771 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031089.2 NP_112351.1 9 - 10 1191 145
GenBank mRNA U55836.1 - 9 - 10 1191 145

View Full Product Details