Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chst15
Entrez Gene ID
286974
Gene Name
carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
Gene Aliases
GalNAc4S6ST, Galnac4s-6st
Chromosome Location
Chr.1: 204243396 - 204328970 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173310.3 NP_775432.3 6 - 7 1510 71
XM_006230430.2 XP_006230492.1 7 - 8 3077 71
XM_006230431.2 XP_006230493.1 7 - 8 3817 71
XM_006230433.2 XP_006230495.1 7 - 8 3312 71
XM_008759868.1 XP_008758090.1 7 - 8 2073 71
GenBank mRNA AB086953.1 - 6 - 7 1510 71

View Full Product Details