Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc36a3
Entrez Gene ID
303148
Gene Name
solute carrier family 36, member 3
Gene Aliases
-
Chromosome Location
Chr.10: 40462752 - 40490083 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024764.1 NP_001019935.1 3 - 4 455 63
XM_006246381.2 XP_006246443.1 3 - 4 498 63
XM_008767685.1 XP_008765907.1 4 - 5 435 63
GenBank mRNA BC097463.1 - 3 - 4 455 63

View Full Product Details