Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbx22
Entrez Gene ID
302369
Gene Name
T-box 22
Gene Aliases
Tbx 22
Chromosome Location
Chr.X: 78731738 - 78782542 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106937.1 NP_001100407.1 4 - 5 494 64
XM_006257159.2 XP_006257221.1 5 - 6 763 64
XM_006257160.2 XP_006257222.1 5 - 6 613 64
XM_006257161.2 XP_006257223.1 5 - 6 760 64
XM_006257162.2 XP_006257224.1 4 - 5 803 64
XM_008773339.1 XP_008771561.1 4 - 5 478 64

View Full Product Details