Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akap12
Entrez Gene ID
83425
Gene Name
A kinase (PRKA) anchor protein 12
Gene Aliases
AKAP12A, AKAP12B, AKAP12G
Chromosome Location
Chr.1: 40816130 - 40906582 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006227847.2 XP_006227909.1 1 - 2 684 81

View Full Product Details