Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aldh5a1
Entrez Gene ID
291133
Gene Name
aldehyde dehydrogenase 5 family, member A1
Gene Aliases
Ssadh
Chromosome Location
Chr.17: 42132085 - 42159413 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022851.1 NP_074042.1 2 - 3 417 59
GenBank mRNA FM032642.1 - 2 - 3 417 59
L34821.1 - 2 - 3 296 59

View Full Product Details