The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047084.1 | NP_001040549.1 | 4 - 5 | 451 | 126 |
XM_008769548.1 | XP_008767770.1 | 4 - 5 | 477 | 126 | |
GenBank mRNA | AY651776.1 | - | 4 - 5 | 451 | 126 |