The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099502.1 | NP_001092972.1 | - | 2016 | 134 |
GenBank mRNA | AF053990.1 | - | - | 228 | 134 |