The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138838.1 | NP_620193.1 | 1 - 1 | 451 | 157 |
GenBank mRNA | M63712.1 | - | 1 - 1 | 312 | 157 |
M72711.1 | - | 1 - 1 | 451 | 157 |