Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-M10-1
Entrez Gene ID
414787
Gene Name
RT1 class I, locus M10, gene 1
Gene Aliases
H2-M10.6
Chromosome Location
Chr.20: 2484136 - 2489529 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008851.1 NP_001008851.1 2 - 3 345 98

View Full Product Details