The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037205.1 | NP_001032282.1 | 11 - 12 | 1382 | 145 |
GenBank mRNA | BC100073.1 | - | 11 - 12 | 1382 | 145 |