Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bex2
Entrez Gene ID
363498
Gene Name
brain expressed X-linked 2
Gene Aliases
EG2RVC
Chromosome Location
Chr.X: 106556838 - 106558366 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077435.1 NP_001070903.1 3 - 3 263 153
GenBank mRNA AY833555.1 - 3 - 3 263 153

View Full Product Details