Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFA2
Entrez Gene ID
853936
Gene Name
Gene Aliases
YKR062W
Chromosome Location
Chr.XI: 559666 - 560652 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179852.1 NP_012988.1 1 - 1 746 99

View Full Product Details