Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOK1
Entrez Gene ID
100233179
Gene Name
docking protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 68494516 - 68497374 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143705.2 NP_001137177.2 5 - 5 1776 93
GenBank mRNA AK348967.1 - 1 - 1 93
FJ438468.1 - 5 - 5 1749 93

View Full Product Details