Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPANK1
Entrez Gene ID
100156389
Gene Name
G-patch domain and ankyrin repeats 1
Gene Aliases
BAT4
Chromosome Location
Chr.7: 23777104 - 23780835 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145383.1 NP_001138855.1 3 - 3 1647 94
XM_005665781.3 XP_005665838.1 - 1596 94
XM_005665782.3 XP_005665839.1 - 1600 94
XM_013977624.2 XP_013833078.1 - 1649 94
XM_021098085.1 XP_020953744.1 - 1808 94
XM_021098086.1 XP_020953745.1 - 2010 94
XM_021098087.1 XP_020953746.1 - 1540 94
XM_021098088.1 XP_020953747.1 - 2166 94
XM_021098089.1 XP_020953748.1 - 1702 94
XM_021098090.1 XP_020953749.1 - 1954 94
XM_021098091.1 XP_020953750.1 - 1914 94
XM_021098092.1 XP_020953751.1 - 1917 94
GenBank mRNA AK235586.1 - 4 - 4 1515 94
AK394180.1 - 3 - 3 1284 94
AK396102.1 - 4 - 4 1514 94
EU282342.2 - 4 - 4 1517 94

View Full Product Details