Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL6
Entrez Gene ID
100517369
Gene Name
methyltransferase like 6
Gene Aliases
-
Chromosome Location
Chr.13: 2638972 - 2654845 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206443.1 NP_001193372.1 4 - 5 1664 102
XM_021068433.1 XP_020924092.1 - 911 102
GenBank mRNA AK233563.1 - 4 - 5 1663 102

View Full Product Details