Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALNT15
Entrez Gene ID
100157572
Gene Name
polypeptide N-acetylgalactosaminyltransferase 15
Gene Aliases
GALNTL2
Chromosome Location
Chr.13: 3329824 - 3371094 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206447.1 NP_001193376.1 5 - 6 1720 74
GenBank mRNA AK351123.1 - 5 - 6 1718 74
AK389431.1 - 5 - 6 1147 74

View Full Product Details