Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGS18
Entrez Gene ID
100511770
Gene Name
regulator of G protein signaling 18
Gene Aliases
-
Chromosome Location
Chr.10: 1520866 - 1539776 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243893.1 NP_001230822.1 1 - 2 331 86
GenBank mRNA AK237063.1 - 1 - 2 331 86
AK391421.1 - 1 - 2 406 86

View Full Product Details