Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
DERL1
Entrez Gene ID
100626802
Gene Name
derlin 1
Gene Aliases
-
Chromosome Location
Chr.4: 16310407 - 16341405 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244479.1 NP_001231408.1 3 - 4 475 91
XM_021088668.1 XP_020944327.1 - 480 91
XM_021088669.1 XP_020944328.1 - 792 91
XM_021088670.1 XP_020944329.1 - 772 91
GenBank mRNA AY610199.1 - 3 - 4 475 91

View Full Product Details