Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdc34
Entrez Gene ID
448013
Gene Name
cell division cycle 34
Gene Aliases
e2-cdc34, ubc3, ube2r1
Chromosome Location
Chr.1: 86544389 - 86552247 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004793.1 NP_001004793.1 2 - 3 424 85
GenBank mRNA BC074529.1 - 2 - 3 424 85
CR760992.2 - 2 - 3 287 85

View Full Product Details