Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
rsph3
Entrez Gene ID
407929
Gene Name
radial spoke head 3 homolog
Gene Aliases
rshl2, rsp3
Chromosome Location
Chr.5: 67068273 - 67093765 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213698.1 NP_998863.1 6 - 7 901 58
XM_012962860.2 XP_012818314.1 6 - 7 876 58
GenBank mRNA BC067913.1 - 6 - 7 901 58

View Full Product Details