Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncbp2
Entrez Gene ID
448671
Gene Name
nuclear cap binding protein subunit 2
Gene Aliases
CBP20
Chromosome Location
Chr.5: 97868455 - 97872205 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006878.1 NP_001006879.1 3 - 4 423 66
GenBank mRNA BC077024.1 - 3 - 4 423 66
CR761328.2 - 3 - 4 392 66

View Full Product Details