Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rps24
Entrez Gene ID
549379
Gene Name
ribosomal protein S24
Gene Aliases
-
Chromosome Location
Chr.7: 56352276 - 56362057 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016625.3 NP_001016625.1 4 - 5 403 78
GenBank mRNA BC135566.1 - 4 - 5 403 78
CR760480.2 - 3 - 4 395 78

View Full Product Details