Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
avd
Entrez Gene ID
100127566
Gene Name
avidin
Gene Aliases
-
Chromosome Location
Chr.1: 167889683 - 167894045 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112934.1 NP_001106405.1 2 - 3 297 82
GenBank mRNA BC152704.1 - 2 - 3 297 82

View Full Product Details