Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
birc7
Entrez Gene ID
100127811
Gene Name
baculoviral IAP repeat containing 7
Gene Aliases
EIAP/XLX, XLX, birc7-a, birc7-b, kiap, livin, ml-iap, mliap, rnf50, xEIAP
Chromosome Location
Chr.Un|NW_016685397: 2479 - 6726 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113122.1 NP_001106593.2 1 - 2 571 79
GenBank mRNA BC155423.1 - 1 - 2 571 79

View Full Product Details