Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccdc59
Entrez Gene ID
100135227
Gene Name
coiled-coil domain containing 59
Gene Aliases
-
Chromosome Location
Chr.3: 43452418 - 43459372 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113924.1 NP_001107396.1 3 - 4 626 99
GenBank mRNA BC157336.1 - 3 - 4 626 99
BC171280.1 - 3 - 4 560 99
BC171282.1 - 3 - 4 560 99
CR855760.2 - 3 - 4 664 99

View Full Product Details