Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ripply3
Entrez Gene ID
100145593
Gene Name
ripply transcriptional repressor 3
Gene Aliases
XRipply3, Xdscr6
Chromosome Location
Chr.2: 8619441 - 8625997 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127005.1 NP_001120477.1 1 - 2 136 77
GenBank mRNA BC075494.1 - 1 - 2 136 77
BC161333.1 - 1 - 2 136 77
CU075356.1 - 1 - 2 146 77

View Full Product Details