Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kif19
Entrez Gene ID
100145702
Gene Name
kinesin family member 19
Gene Aliases
-
Chromosome Location
Chr.10: 25044633 - 25094873 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012959642.2 XP_012815096.1 8 - 9 1434 61
GenBank mRNA BC161478.1 - - 1123 61

View Full Product Details