Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
exosc1
Entrez Gene ID
100170616
Gene Name
exosome component 1
Gene Aliases
-
Chromosome Location
Chr.7: 24625098 - 24629813 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130379.1 NP_001123851.1 4 - 5 332 100
GenBank mRNA BC167725.1 - 4 - 5 332 100
CR942534.2 - 4 - 5 323 100

View Full Product Details