Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aldh5a1
Entrez Gene ID
100216127
Gene Name
aldehyde dehydrogenase 5 family member A1
Gene Aliases
-
Chromosome Location
Chr.6: 31482856 - 31493342 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142106.1 NP_001135578.1 8 - 9 1477 75
XM_018094569.1 XP_017950058.1 7 - 8 1395 75
GenBank mRNA BC124570.1 - 8 - 9 1826 75
BC168085.1 - 8 - 9 1477 75

View Full Product Details