Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ANT1
Entrez Gene ID
542386
Gene Name
adenine nucleotide translocator
Gene Aliases
MANT1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111961.1 NP_001105431.1 - 951 91
GenBank mRNA BT043116.1 - - 954 91
EU952880.1 - - 971 91
X57556.1 - - 951 91

View Full Product Details