Gene Transcripts
- Gene Symbol
- DGCR10
- Entrez Gene ID
- 26222
- Gene Name
- DiGeorge syndrome critical region gene 10 (non-protein coding)
- Gene Aliases
- DGS-B
- Chromosome Location
- Chr. 22: 19022624 - 19023550 on Build GRCh38
- UniGene ID
- Hs.681383
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026651.1 |
|
1 |
244 |
|
GenBank mRNA |
BU584104.1 |
- |
1 |
243 |
|
L77559.1 |
- |
1 |
83 |
|
View Full Product Details