Gene Transcripts

Gene Symbol
FAM167A-AS1
Entrez Gene ID
83656
Gene Name
FAM167A antisense RNA 1
Gene Aliases
C8orf12
Chromosome Location
Chr. 8: 11368402 - 11438657 on Build GRCh38
UniGene ID
Hs.583896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026814.1 2 220
GenBank mRNA AJ301563.1 - 2 220


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000533578.4 - 1, 2 220 Chr. 8 - 11433920 - 11438658 [+]

View Full Product Details