Gene Transcripts
- Gene Symbol
- CYP4F35P
- Entrez Gene ID
- 284233
- Gene Name
- cytochrome P450 family 4 subfamily F member 35, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 18: 14337423 - 14342524 on Build GRCh38
- UniGene ID
- Hs.607579
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026756.1 |
|
4 |
2016 |
|
GenBank mRNA |
BC037172.1 |
- |
4 |
2016 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ899076.1 |
- |
1 |
705 |
|
Chr. 21 - 13843806 - 13844696 [-] |
View Full Product Details