Gene Transcripts

Gene Symbol
ZC3H14
Entrez Gene ID
79882
Gene Name
zinc finger CCCH-type containing 14
Gene Aliases
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location
Chr. 14: 88562909 - 88614720 on Build GRCh38
UniGene ID
Hs.684046
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011537162.2 XP_011535464.1 10 6051
GenBank mRNA AL832086.1 - 1 1892

View Full Product Details