Gene Transcripts

Gene Symbol
DIO3OS
Entrez Gene ID
64150
Gene Name
DIO3 opposite strand/antisense RNA (head to head)
Gene Aliases
C14orf134, DIO3-AS1, DIO3-OS, NCRNA00041
Chromosome Location
Chr. 14: 101552223 - 101555676 on Build GRCh38
UniGene ID
Hs.525597
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC037351.1 - 1 2267
BC065701.1 - 3 1041
BC111049.1 - 2 858

View Full Product Details