Gene Transcripts
- Gene Symbol
- SECISBP2
- Entrez Gene ID
- 79048
- Gene Name
- SECIS binding protein 2
- Gene Aliases
- SBP2
- Chromosome Location
- Chr. 9: 89318473 - 89367117 on Build GRCh38
- UniGene ID
- Hs.59804
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127655.1 |
- |
1 |
2016 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB282642.1 |
- |
1 |
152 |
|
Chr. 9 - 89354986 - 89355536 [+] |
View Full Product Details