Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270407.1 | NP_001257336.1 | 9 | 3771 | |
NM_021219.3 | NP_067042.1 | 10 | 3879 | ||
NR_072999.1 | 10 | 3875 | |||
GenBank mRNA | AK056079.1 | - | 1 | 1962 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DR423676.1 | - | 1 | 761 | Chr. 21 - 25716354 - 25717164 [+] |