Gene Transcripts
- Gene Symbol
- Dnajb4
- Entrez Gene ID
- 67035
- Gene Name
- DnaJ (Hsp40) homolog, subfamily B, member 4
- Gene Aliases
- 1700029A20Rik, 2010306G19Rik, 5730460G06Rik
- Chromosome Location
- Chr. 3: 152183871 - 152210333 on Build GRCm38
- UniGene ID
- Mm.458538
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077587.1 |
- |
1 |
993 |
|
AK136335.1 |
- |
1 |
387 |
|
View Full Product Details