Gene Transcripts
- Gene Symbol
- Grm2
- Entrez Gene ID
- 108068
- Gene Name
- glutamate receptor, metabotropic 2
- Gene Aliases
- 4930441L02Rik, Gprc1b, mGluR2, mGluR7
- Chromosome Location
- Chr. 9: 106644534 - 106656109 on Build GRCm38
- UniGene ID
- Mm.410822
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015353.1 |
- |
|
1068 |
|
View Full Product Details