Gene Transcripts

Gene Symbol
SFTA3
Entrez Gene ID
253970
Gene Name
surfactant associated 3
Gene Aliases
NANCI, SFTPH, SP-H
Chromosome Location
Chr. 14: 36473288 - 36513785 on Build GRCh38
UniGene ID
Hs.509165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY102071.1 - 5 1052


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000521945.1 - 5 1052 Chr. 14 - 36473207 - 36519521 [-]

View Full Product Details