Gene Transcripts
- Gene Symbol
- DIO3OS
- Entrez Gene ID
- 64150
- Gene Name
- DIO3 opposite strand/antisense RNA (head to head)
- Gene Aliases
- C14orf134, DIO3-AS1, DIO3-OS, NCRNA00041
- Chromosome Location
- Chr. 14: 101552223 - 101555676 on Build GRCh38
- UniGene ID
- Hs.525597
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000555174.5 |
- |
2 |
1642 |
|
Chr. 14 - 101557304 - 101560422 [-] |
GENCODE |
ENST00000554694.1 |
- |
3 |
810 |
|
Chr. 14 - 101557308 - 101560403 [-] |
GENCODE |
ENST00000554441.5 |
- |
4 |
1008 |
|
Chr. 14 - 101557304 - 101560375 [-] |
GENCODE |
ENST00000557661.1 |
- |
3 |
1877 |
|
Chr. 14 - 101557304 - 101560431 [-] |
GENCODE |
ENST00000557532.5 |
- |
4 |
656 |
|
Chr. 14 - 101557308 - 101560403 [-] |
View Full Product Details